accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2769-64-4

IUPAC Name: 1-isocyanobutane
Synonyms: N-butyl isocyanide n-butyl isocyanide n-butylisocyanide butyl isocyanide
Molecular Weight (g/mol): 83.13
Molecular Formula: C5H9N
InChi Key: FSBLVBBRXSCOKU-UHFFFAOYSA-N
SMILES: CCCC[N+]#[C-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results