accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2769-71-3

CH3H3CNC

CAS RN 2769-71-3

IUPAC Name: 2-isocyano-1,3-dimethylbenzene
Synonyms: 2-isocyano-1,3-dimethylbenzene
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: DNJLFZHMJDSJFN-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1[N+]#[C-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results