accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 277319-62-7

IUPAC Name: 4-amino-N'-hydroxybenzene-1-carboximidamide
Synonyms: 4-amino-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 151.17
Molecular Formula: C7H9N3O
InChi Key: CNFNMMJKXWOLPY-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results