accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OPOOOH3CCH3H3COH3C

CAS RN 27784-76-5

IUPAC Name: tert-butyl 2-(diethoxyphosphoryl)acetate
Synonyms: tert-butyl 2-(diethoxyphosphoryl)acetate
Molecular Weight (g/mol): 252.25
Molecular Formula: C10H21O5P
InChi Key: NFEGNISFSSLEGU-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC(=O)OC(C)(C)C)OCC
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)