accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27813-02-1

IUPAC Name: 1-hydroxypropan-2-yl 2-methylprop-2-enoate
Synonyms: hydroxyisopropyl methacrylate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: ZMARGGQEAJXRFP-UHFFFAOYNA-N
SMILES: CC(CO)OC(=O)C(C)=C
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results