accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNOO

CAS RN 27828-71-3

IUPAC Name: 5-hydroxypyridine-3-carboxylate
Synonyms: 5-hydroxypyridine-3-carboxylate
Molecular Weight (g/mol): 138.1
Molecular Formula: C6H4NO3
InChi Key: ATTDCVLRGFEHEO-UHFFFAOYSA-M
SMILES: OC1=CN=CC(=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results