accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2785-89-9

H3COHOH3C

CAS RN 2785-89-9

IUPAC Name: 4-ethyl-2-methoxyphenol
Synonyms: 4-ethylguaiacol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: CHWNEIVBYREQRF-UHFFFAOYSA-N
SMILES: CCC1=CC=C(O)C(OC)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)