accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2788-86-5

IUPAC Name: 2-(4-chlorophenyl)oxirane
Synonyms: 2-(4-chlorophenyl)oxirane
Molecular Weight (g/mol): 154.59
Molecular Formula: C8H7ClO
InChi Key: IBWLXNDOMYKTAD-UHFFFAOYNA-N
SMILES: ClC1=CC=C(C=C1)C1CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results