accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH2OHOFFF

CAS RN 2792-72-5

IUPAC Name: 2-amino-5,5,5-trifluoro-4-methylpentanoic acid
Synonyms: 2-amino-5,5,5-trifluoro-4-methylpentanoic acid
Molecular Weight (g/mol): 185.15
Molecular Formula: C6H10F3NO2
InChi Key: XFGVJLGVINCWDP-UHFFFAOYNA-N
SMILES: CC(CC(N)C(O)=O)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)