accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFClBr

CAS RN 279252-26-5

IUPAC Name: 1-(bromomethyl)-2-chloro-4-(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-2-chloro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 273.48
Molecular Formula: C8H5BrClF3
InChi Key: AIZUUFYFLNVTQX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results