accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OH

CAS RN 27975-78-6

IUPAC Name: 1-phenylpent-1-yn-3-ol
Synonyms: 1-phenylpent-1-yn-3-ol
Molecular Weight (g/mol): 160.22
Molecular Formula: C11H12O
InChi Key: QWCMSASONHVIHV-UHFFFAOYNA-N
SMILES: CCC(O)C#CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results