accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27996-87-8

IUPAC Name: 2-fluoro-5-nitrobenzaldehyde
Synonyms: 2-fluoro-5-nitrobenzaldehyde
Molecular Weight (g/mol): 169.11
Molecular Formula: C7H4FNO3
InChi Key: VVXFDFQEIRGULC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C(C=O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results