accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHCl

CAS RN 28034-99-3

IUPAC Name: 4'-chloro-[1,1'-biphenyl]-4-ol
Synonyms: 4-chloro-4'-biphenylol
Molecular Weight (g/mol): 204.65
Molecular Formula: C12H9ClO
InChi Key: ICVFJPSNAUMFCW-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)