accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 28080-54-8

IUPAC Name: 2-iodo-5-nitropyridine
Synonyms: 2-iodo-5-nitropyridine
Molecular Weight (g/mol): 250
Molecular Formula: C5H3IN2O2
InChi Key: SJXWHBQWFBHASX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results