accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3N(S)HO(S)OH

CAS RN 28143-91-1

IUPAC Name: (1S,2S)-1,3-dihydroxy-1-phenylpropan-2-aminium
Synonyms: (1S,2S)-1,3-dihydroxy-1-phenylpropan-2-aminium
Molecular Weight (g/mol): 168.22
Molecular Formula: C9H14NO2
InChi Key: JUCGVCVPNPBJIG-IUCAKERBSA-O
SMILES: [NH3+][C@@H](CO)[C@@H](O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)