accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 28169-46-2

IUPAC Name: 2-methyl-3,5-dinitrobenzoic acid
Synonyms: 2-methyl-3,5-dinitrobenzoic acid
Molecular Weight (g/mol): 226.14
Molecular Formula: C8H6N2O6
InChi Key: CDVNZMKTJIBBBV-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results