accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 28170-13-0

OSHK

CAS RN 28170-13-0

IUPAC Name: benzenecarbothioic S-acid potassium
Synonyms: benzenecarbothioic acid potassium
Molecular Weight (g/mol): 177.28
Molecular Formula: C7H6KOS
InChi Key: LSBMSRUYNGEWKI-UHFFFAOYSA-N
SMILES: [K].SC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results