accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 28188-41-2

BrN

CAS RN 28188-41-2

IUPAC Name: 3-(bromomethyl)benzonitrile
Synonyms: 3-(bromomethyl)benzonitrile
Molecular Weight (g/mol): 196.05
Molecular Formula: C8H6BrN
InChi Key: CVKOOKPNCVYHNY-UHFFFAOYSA-N
SMILES: BrCC1=CC=CC(=C1)C#N
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)