accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2824-57-9

IUPAC Name: methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
Synonyms: methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
Molecular Weight (g/mol): 260.29
Molecular Formula: C14H16N2O3
InChi Key: XZECNVJPYDPBAM-ZDUSSCGKSA-N
SMILES: COC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)