accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOOOCH3OOH3COOCH3OOCH3

CAS RN 28244-99-7

IUPAC Name: [3,4,5-tris(acetyloxy)-6-[(4-methylphenyl)sulfanyl]oxan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)-6-[(4-methylphenyl)sulfanyl]oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 454.49
Molecular Formula: C21H26O9S
InChi Key: XRFOIRMXQHXTRL-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(SC2=CC=C(C)C=C2)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results