CAS RN 28272-96-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Ethenylbenzaldehyde, TRC
CAS: 28272-96-0 Molecular Formula: C9H8O Molecular Weight (g/mol): 132.16 Synonym: 2-Formylstyrene,2-Vinylbenzaldehyde,o-Formylstyrene,o-Vinylbenzaldehyde SMILES: C=Cc1ccccc1C=O