accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CMgClH3CCH3

CAS RN 28276-08-6

IUPAC Name: magnesium(2+) 2-methylbutan-2-ide chloride
Synonyms: magnesium(2+) 2-methylbutan-2-ide chloride
Molecular Weight (g/mol): 130.9
Molecular Formula: C5H11ClMg
InChi Key: FKKWHMOEKFXMPU-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].CC[C-](C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results