CAS RN 28300-74-5
IUPAC Name:
dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate
Synonyms:
antimony potassium tartrate
Molecular Weight (g/mol):
667.87
Molecular Formula:
C8H10K2O15Sb2
InChi Key:
WBTCZEPSIIFINA-UHFFFAOYNA-J
SMILES:
O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
1
–
3
of
3
results
1
Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%
CAS: 28300-74-5 Molecular Formula: C8H4K2O12Sb2·3H2O MDL Number: MFCD00148863 Synonym: Potassium antimonyl tartrate
2
Potassium antimonyl tartrate trihydrate, 99+%
CAS: 28300-74-5 Molecular Formula: C8H10K2O15Sb2 Molecular Weight (g/mol): 667.87 MDL Number: MFCD00167056 Synonym: Antimonyl potassium tartrate sesquihydrate