CAS RN 28328-53-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-Nitro-4-phenoxy-5-sulfamoylbenzoic Acid, TRC
CAS: 28328-53-2 Molecular Formula: C13 H10 N2 O7 S Molecular Weight (g/mol): 338.29 Synonym: Benzoic acid, 3-nitro-4-phenoxy-5-sulfamoyl- (8CI),3-Nitro-4-phenoxy-5-sulfamoylbenzoic acid IUPAC Name: 3-nitro-4-phenoxy-5-sulfamoylbenzoic acid SMILES: NS(=O)(=O)c1cc(cc(c1Oc2ccccc2)[N+](=O)[O-])C(=O)O
2
3-Nitro-4-phenoxy-5-sulfamoylbenzoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.