CAS RN 28328-54-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Desbutyl Bumetanide, TRC
CAS: 28328-54-3 Molecular Formula: C13 H12 N2 O5 S Molecular Weight (g/mol): 308.31 Synonym: Benzoic acid, 3-amino-5-(aminosulfonyl)-4-phenoxy-,Benzoic acid, 3-amino-4-phenoxy-5-sulfamoyl- (8CI),3-Amino-4-phenoxy-5-sulfamoylbenzoic acid,Desbutylbumetanide,PF 1578 IUPAC Name: 3-amino-4-phenoxy-5-sulfamoylbenzoic acid SMILES: Nc1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)O
2
3-Amino-4-phenoxy-5-sulfamoylbenzoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.