accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2835-98-5

CH3NH2HO

CAS RN 2835-98-5

IUPAC Name: 2-amino-5-methylphenol
Synonyms: 6-amino-M-cresol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: HCPJEHJGFKWRFM-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results