accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHOOH

CAS RN 28393-02-4

IUPAC Name: docosa-10,12-diynedioic acid
Synonyms: docosa-10,12-diynedioic acid
Molecular Weight (g/mol): 362.51
Molecular Formula: C22H34O4
InChi Key: XCUAGNLOPZWTEH-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCC#CC#CCCCCCCCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results