accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 28395-03-1

IUPAC Name: 3-(butylamino)-4-phenoxy-5-sulfamoylbenzoic acid
Synonyms: bumetanide bumethanide segurex yurinex bumetanide astrazeneca brand fordiuran grossmann brand of bumetanide farmacusi brand of bumetanide senosiain brand of bumetanide atlantis brand of bumetanide
Molecular Weight (g/mol): 364.42
Molecular Formula: C17H20N2O5S
InChi Key: MAEIEVLCKWDQJH-UHFFFAOYSA-N
SMILES: CCCCNC1=CC(=CC(=C1OC1=CC=CC=C1)S(N)(=O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)