accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COH2NOO

CAS RN 2840-26-8

IUPAC Name: 3-amino-4-methoxybenzoate
Synonyms: 3-amino-4-methoxybenzoate
Molecular Weight (g/mol): 166.16
Molecular Formula: C8H8NO3
InChi Key: FDGAEAYZQQCBRN-UHFFFAOYSA-M
SMILES: COC1=CC=C(C=C1N)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results