accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 28425-04-9

IUPAC Name: dichloropalladium; bis(triethylphosphane)
Synonyms: palladium chloride; bis(triethylphosphine)
Molecular Weight (g/mol): 413.64
Molecular Formula: C12H30Cl2P2Pd
InChi Key: ULYNIEUXPCUIEL-UHFFFAOYSA-L
SMILES: Cl[Pd]Cl.CCP(CC)CC.CCP(CC)CC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results