accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 28443-50-7

IUPAC Name: 2-amino-5-chlorophenol
Synonyms: 2-amino-5-chlorophenol
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: FZCQMIRJCGWWCL-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)