accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 284493-53-4

CH3O(S)OONHOOCH3CH3H3C

CAS RN 284493-53-4

IUPAC Name: (3S)-3-{[(tert-butoxy)carbonyl]amino}-3-(3-methoxyphenyl)propanoate
Synonyms: (3S)-3-[(tert-butoxycarbonyl)amino]-3-(3-methoxyphenyl)propanoate
Molecular Weight (g/mol): 294.33
Molecular Formula: C15H20NO5
InChi Key: DNXONWHJJNOKEU-LBPRGKRZSA-M
SMILES: COC1=CC=CC(=C1)[C@H](CC([O-])=O)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results