accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 284493-59-0

CH3H3CCH3OONHOOHF

CAS RN 284493-59-0

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}-3-(3-fluorophenyl)propanoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]-3-(3-fluorophenyl)propanoic acid
Molecular Weight (g/mol): 283.3
Molecular Formula: C14H18FNO4
InChi Key: IQPQPXUDXQDVMK-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(O)=O)C1=CC(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results