accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SNCO

CAS RN 28479-19-8

IUPAC Name: 1-isocyanato-3-(methylsulfanyl)benzene
Synonyms: 1-isocyanato-3-(methylsulfanyl)benzene
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: BKJABLMNBSVKCV-UHFFFAOYSA-N
SMILES: CSC1=CC=CC(=C1)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results