accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 28485-17-8

H3COONHOHNO

CAS RN 28485-17-8

IUPAC Name: ethyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Synonyms: ethyl 2,4-dioxo-1,3-dihydropyrimidine-5-carboxylate
Molecular Weight (g/mol): 184.15
Molecular Formula: C7H8N2O4
InChi Key: MKNYHTGOVKPZMU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CNC(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)