accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OH3COCu

CAS RN 2850-65-9

IUPAC Name: copper(2+) diethanolate
Synonyms: copper(2+) bis(ethoxide)
Molecular Weight (g/mol): 153.67
Molecular Formula: C4H10CuO2
InChi Key: CRCKGIUJMFFISH-UHFFFAOYSA-N
SMILES: [Cu++].CC[O-].CC[O-]
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results