CAS RN 285570-60-7
IUPAC Name:
3-{2-[4-carboxy-2-(3-carboxy-2-acetamidopropanamido)butanamido]-3-methylbutanamido}-5-chloro-4-oxopentanoic acid
Synonyms:
3-{2-[4-carboxy-2-(3-carboxy-2-acetamidopropanamido)butanamido]-3-methylbutanamido}-5-chloro-4-oxopentanoic acid
Molecular Weight (g/mol):
550.95
Molecular Formula:
C21H31ClN4O11
InChi Key:
ATNOUPFYBMVFLD-UHFFFAOYNA-N
SMILES:
CC(C)C(NC(=O)C(CCC(O)=O)NC(=O)C(CC(O)=O)NC(C)=O)C(=O)NC(CC(O)=O)C(=O)CCl
1
–
1
of
1
results
1
Thermo Scientific Chemicals Caspase-3 Inhibitor III
CAS: 285570-60-7 Molecular Formula: C21H31ClN4O11 Synonym: Ac-DEVD-CMK; N-Acetyl-Asp-Glu-Val-Asp chloromethyl ketone