accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOBr

CAS RN 28598-81-4

IUPAC Name: ethyl 9-bromononanoate
Synonyms: ethyl 9-bromononanoate
Molecular Weight (g/mol): 265.19
Molecular Formula: C11H21BrO2
InChi Key: KXZBPLOPBKIUTC-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCCCCBr
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)