accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O

CAS RN 286-20-4

IUPAC Name: 7-oxabicyclo[4.1.0]heptane
Synonyms: 7-oxabicyclo 4.1.0 heptane
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: ZWAJLVLEBYIOTI-UHFFFAOYNA-N
SMILES: C1CCC2OC2C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)