accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BFFFFNN

CAS RN 286014-38-8

IUPAC Name: 1,3-dicyclohexyl-3H-1λ⁵-imidazol-1-ylium; tetrafluoroboranuide
Synonyms: 1,3-dicyclohexyl-1λ⁵-imidazol-1-ylium tetrafluoroborate
Molecular Weight (g/mol): 320.18
Molecular Formula: C15H25BF4N2
InChi Key: CQHXJIHJFMBBQA-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results