accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2863-98-1

IUPAC Name: 4-hydrazinylbenzonitrile
Synonyms: 4-hydrazinylbenzonitrile
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: DZUUSHCOMPROCJ-UHFFFAOYSA-N
SMILES: NNC1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (7)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (4)