CAS RN 287-92-3
Cyclopentane, 97%, extra pure
CAS: 287-92-3 Molecular Formula: C5H10 Molecular Weight (g/mol): 70.15 MDL Number: MFCD00001356 InChI Key: RGSFGYAAUTVSQA-UHFFFAOYSA-N Synonym: pentamethylene,ciclopentano,cyclopentan,zyklopentan,unii-t86pb90rnu,hsdb 62,t86pb90rnu,cyclopentanes,cyclopentyl group,hydrocarbons, cyclic c5 and c6 PubChem CID: 9253 ChEBI: CHEBI:23492 IUPAC Name: cyclopentane SMILES: C1CCCC1
Cyclopentane, HPLC Grade
CAS: 287-92-3 Molecular Formula: C5H10 Molecular Weight (g/mol): 70.135 MDL Number: MFCD00001356 InChI Key: RGSFGYAAUTVSQA-UHFFFAOYSA-N Synonym: pentamethylene,ciclopentano,cyclopentan,zyklopentan,unii-t86pb90rnu,hsdb 62,t86pb90rnu,cyclopentanes,cyclopentyl group,hydrocarbons, cyclic c5 and c6 PubChem CID: 9253 ChEBI: CHEBI:23492 IUPAC Name: cyclopentane SMILES: C1CCCC1
Cyclopentane, 95+%, pure
CAS: 287-92-3 Molecular Formula: C5H10 Molecular Weight (g/mol): 70.15 MDL Number: MFCD00001356 InChI Key: RGSFGYAAUTVSQA-UHFFFAOYSA-N Synonym: pentamethylene,ciclopentano,cyclopentan,zyklopentan,unii-t86pb90rnu,hsdb 62,t86pb90rnu,cyclopentanes,cyclopentyl group,hydrocarbons, cyclic c5 and c6 PubChem CID: 9253 ChEBI: CHEBI:23492 IUPAC Name: cyclopentane SMILES: C1CCCC1
Cyclopentane, 95%
CAS: 287-92-3 Molecular Formula: C5H10 Molecular Weight (g/mol): 70.135 MDL Number: MFCD00001356 InChI Key: RGSFGYAAUTVSQA-UHFFFAOYSA-N Synonym: pentamethylene,ciclopentano,cyclopentan,zyklopentan,unii-t86pb90rnu,hsdb 62,t86pb90rnu,cyclopentanes,cyclopentyl group,hydrocarbons, cyclic c5 and c6 PubChem CID: 9253 ChEBI: CHEBI:23492 IUPAC Name: cyclopentane SMILES: C1CCCC1
ASTM Method D5191 Vapor Pressure - 68.3kPa (9.91 psi), Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Cyclopentane, Reagent Grade, 98%, Merck
CAS: 287-92-3 Molecular Formula: C5H10 Molecular Weight (g/mol): 70.135 MDL Number: MFCD00001356 InChI Key: RGSFGYAAUTVSQA-UHFFFAOYSA-N Synonym: pentamethylene,ciclopentano,cyclopentan,zyklopentan,unii-t86pb90rnu,hsdb 62,t86pb90rnu,cyclopentanes,cyclopentyl group,hydrocarbons, cyclic c5 and c6 PubChem CID: 9253 ChEBI: CHEBI:23492 IUPAC Name: cyclopentane SMILES: C1CCCC1