accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFFFF

CAS RN 287172-66-1

IUPAC Name: 1-(trifluoromethyl)-2-{2-[2-(trifluoromethyl)phenyl]ethyl}benzene
Synonyms: 1-(trifluoromethyl)-2-{2-[2-(trifluoromethyl)phenyl]ethyl}benzene
Molecular Weight (g/mol): 318.26
Molecular Formula: C16H12F6
InChi Key: OBMDBKNYJGWWJX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1CCC1=CC=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results