accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HHClClH2NHNHNNH2NH

CAS RN 28718-90-3

IUPAC Name: dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride
Synonyms: DAPI
Molecular Weight (g/mol): 350.25
Molecular Formula: C16H17Cl2N5
InChi Key: FPNZBYLXNYPRLR-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)