accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOCl

CAS RN 287197-95-9

IUPAC Name: 2-(chloromethyl)-5-(4-methylphenyl)-1,3,4-oxadiazole
Synonyms: 2-(chloromethyl)-5-(4-methylphenyl)-1,3,4-oxadiazole
Molecular Weight (g/mol): 208.65
Molecular Formula: C10H9ClN2O
InChi Key: JDAULQMOFMANSL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=NN=C(CCl)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results