accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBOHOH

CAS RN 28741-08-4

IUPAC Name: octylboronic acid
Synonyms: octylboronic acid
Molecular Weight (g/mol): 158.05
Molecular Formula: C8H19BO2
InChi Key: GKFRVXOKPXCXAK-UHFFFAOYSA-N
SMILES: CCCCCCCCB(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results