accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3FF

CAS RN 28781-85-3

IUPAC Name: ethyl 2,2-difluoropropanoate
Synonyms: ethyl 2,2-difluoropropanoate
Molecular Weight (g/mol): 138.11
Molecular Formula: C5H8F2O2
InChi Key: ISVLDAIKRGXNCZ-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results