accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2879-20-1

CH3OOO

CAS RN 2879-20-1

IUPAC Name: 1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-one
Synonyms: 1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: HGVWMTAIIYNQSI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2OCCOC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)