accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNNBr

CAS RN 287922-71-8

IUPAC Name: 4-bromo-1-methyl-1H-pyrazole-3-carbonitrile
Synonyms: 4-bromo-1-methylpyrazole-3-carbonitrile
Molecular Weight (g/mol): 186.01
Molecular Formula: C5H4BrN3
InChi Key: NTTLCOOFUGHMJS-UHFFFAOYSA-N
SMILES: CN1C=C(Br)C(=N1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results