CAS RN 2881-99-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Cyanophenylhydrazine Hydrochloride (contains ~15% inorganics), TRC
CAS: 2881-99-4 Molecular Formula: C7H8ClN3 Molecular Weight (g/mol): 169.61 IUPAC Name: 3-hydrazinylbenzonitrile;hydrochloride SMILES: Cl.NNc1cccc(c1)C#N